4-Nitro-1,8-naphthalic anhydride 95%

Supplier: Ambeed
Warning

Synonyms: 6-Nitrobenzo[de]isochromene-1,3-dione

A507979-10G A507979-1G A507979-250MG A507979-25G A507979-5G
76726-094EA 346.92 USD
76726-094 76726-096 76726-098 76726-100 76726-102
4-Nitro-1,8-naphthalic anhydride 95%
4-Nitro-1,8-naphthalic anhydride

Precaución: for Research Use Only

Formula: C₁₂H₅NO₅
MW: 243.1718 g/mol
MDL Number: MFCD00013446
CAS Number: 6642-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR