4-((3-Methylbut-2-en-1-yl)oxy)benzaldehyde 95%

Supplier: Ambeed
Warning

A507274-1G A507274-250MG
76726-082EA 152.77 USD
76726-082 76726-084
4-((3-Methylbut-2-en-1-yl)oxy)benzaldehyde 95%
4-((3-Methylbut-2-en-1-yl)oxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₂
MW: 190.23 g/mol
MDL Number: MFCD06797957
CAS Number: 28090-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR