(Methylamino)acetaldehyde dimethylacetal 98%

Supplier: Ambeed
Danger

A503993-25G A503993-5G
76726-054EA 8.49 USD
76726-054 76726-056 76726-058
(Methylamino)acetaldehyde dimethylacetal 98%
(Methylamino)acetaldehyde dimethylacetal

Precaución: for Research Use Only

Formula: C₅H₁₃NO₂
MDL Number: MFCD00008485
CAS Number: 122-07-6
UN: 1989
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR