6-Aminobenzo[de]isochromene-1,3-dione 95%

Supplier: Ambeed
Warning

A502980-100MG A502980-1G A502980-250MG A502980-5G
76726-030EA 20.16 USD
76726-030 76726-032 76726-034 76726-036
6-Aminobenzo[de]isochromene-1,3-dione 95%
6-Aminobenzo[de]isochromene-1,3-dione

Precaución: for Research Use Only

Formula: C₁₂H₇NO₃
MW: 213.19 g/mol
MDL Number: MFCD00009776
CAS Number: 6492-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR