5-Methylphenazinium methyl sulfate 97%

Supplier: Ambeed
Warning

A502765-250MG A502765-50MG A502765-5G A502765-1G
76726-024EA 7 USD
76726-024 76726-026 76726-028 77602-166
5-Methylphenazinium methyl sulfate 97%
5-Methylphenazinium methyl sulphate

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O₄S
MDL Number: MFCD00011923
CAS Number: 299-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR