Trimethoxy[3-(phenylamino)propyl]silane 98%

Supplier: Ambeed
Danger

Synonyms: N-[3-(Trimethoxysilyl)propyl]aniline

A497485-25G A497485-5G
76726-016EA 17 USD
76726-016 77880-081
Trimethoxy[3-(phenylamino)propyl]silane 98%
Trimethoxy[3-(phenylamino)propyl]silane

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₃Si
MDL Number: MFCD00053673
CAS Number: 3068-76-6
UN: 3267
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR