Methyl-3-(1,3-oxazol-5-yl)benzoate 97%

Supplier: Ambeed
Warning

A497212-100MG A497212-1G
76726-012EA 39.26 USD
76726-012 76726-014
Methyl-3-(1,3-oxazol-5-yl)benzoate 97%
Methyl-3-(1,3-oxazol-5-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MDL Number: MFCD06797482
CAS Number: 850375-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR