Dipotassium hydrogen orthophosphate trihydrate 99%

Supplier: Ambeed
Warning

A495981-1000G A495981-100G A495981-500G
76726-006EA 10 USD
76726-006 76726-008 76726-010
Dipotassium hydrogen orthophosphate trihydrate 99%
Dipotassium hydrogen orthophosphate trihydrate

Precaución: for Research Use Only

Formula: K₂HPO₄·3H₂O
MDL Number: MFCD00149926
CAS Number: 16788-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR