Sorbitan tristearate 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
Span 65, (2R,3R,4S)-2-((R)-2-Hydroxy-1-(stearoyloxy)ethyl)tetrahydrofuran-3,4-diyl distearate
A495916-1000G
A495916-100G
A495916-25G
A495916-500G
76725-998EA
26
USD
76725-998
76726-000
76726-002
76726-004
Sorbitan tristearate 98+%
Sorbitan tristearate
Precaución: for Research Use Only
Formula:
C₆₀H₁₁₄O₈ |
MDL Number:
MFCD00151165 CAS Number: 26658-19-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...