N-tert-Butyl-3-nitrobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A495244-100MG A495244-1G A495244-250MG A495244-5G
76725-990EA 7 USD
76725-990 76725-992 76725-994 76725-996
N-tert-Butyl-3-nitrobenzenesulfonamide 98%
N-tert-Butyl-3-nitrobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₄S
MDL Number: MFCD01985492
CAS Number: 424818-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR