Methyl laurate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
A491786-25G
A491786-500G
A491786-1000G
A491786-100G
76725-974EA
5.31
USD
76725-974
77602-124
77669-206
77669-208
Methyl laurate 95%
Methyl laurate
Precaución: for Research Use Only
Formula:
C₁₃H₂₆O₂ MW: 214.35 g/mol |
MDL Number:
MFCD00008966 CAS Number: 111-82-0 UN: 3082 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...