Potassium ((diethylamino)methyl)trifluoroborate 95%

Supplier: Ambeed
Warning

A491615-1G A491615-100MG
76725-972EA 113 USD
76725-972 77880-071
Potassium ((diethylamino)methyl)trifluoroborate 95%
Potassium ((diethylamino)methyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₅H₁₂BF₃KN
MW: 193.06 g/mol
MDL Number: MFCD11052700
CAS Number: 936329-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR