Diisobutyl perylenedicarboxylate (mixture of regioisomers) 95%

Supplier: Ambeed
Warning

Synonyms: Perylenedicarboxylic acid Diisobutyl ester

A489789-10G A489789-25G A489789-5G
76725-958EA 9.55 USD
76725-958 76725-960 76725-962
Diisobutyl perylenedicarboxylate (mixture of regioisomers) 95%
Diisobutyl perylenedicarboxylate

Precaución: for Research Use Only

Formula: C₃₀H₂₈O₄
MW: 452.55 g/mol
MDL Number: MFCD00191681
CAS Number: 2744-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR