2-(4-((tert-Butoxycarbonyl)amino)-2-nitrophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A488523-100MG A488523-1G A488523-250MG
76725-952EA 39 USD
76725-952 76725-954 76725-956
2-(4-((tert-Butoxycarbonyl)amino)-2-nitrophenyl)acetic acid 95%
2-(4-((tert-Butoxycarbonyl)amino)-2-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₆
MDL Number: MFCD08236844
CAS Number: 512180-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR