Copper(II) phthalocyanine 98%+ (dye content)
Supplier: Ambeed
|
Warning
|
Synonyms:
29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15
A486818-100G
A486818-25G
A486818-500G
76725-942EA
46
USD
76725-942
76725-944
76725-946
Copper(II) phthalocyanine 98%+ (dye content)
Copper(II) phthalocyanine
Precaución: for Research Use Only
Formula:
C₃₂H₁₆CuN₈ MW: 576.08 g/mol Melting Pt: 480 |
MDL Number:
MFCD00010719 CAS Number: 147-14-8 Merck Index: 12,02584 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...