N-Ethyl-1,3-propanediamine 97%

Supplier: Ambeed
Danger

Synonyms: N1-Ethylpropane-1,3-diamine

A484421-1G A484421-5G
76725-920EA 23.34 USD
76725-920 76725-922
N-Ethyl-1,3-propanediamine 97%
N-Ethyl-1,3-propanediamine

Precaución: for Research Use Only

Formula: C₅H₁₄N₂
MW: 102.1781 g/mol
MDL Number: MFCD00014829
CAS Number: 10563-23-2
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR