Quinuclidine 98%

Supplier: Ambeed
Danger

Synonyms: 1-Azabicyclo[2.2.2]octane, ABCO

A482782-1G A482782-25G A482782-5G
76725-904EA 42.44 USD
76725-904 76725-906 76725-908
Quinuclidine 98%
Quinuclidine

Precaución: for Research Use Only

Formula: C₇H₁₃N
MW: 111.19 g/mol
MDL Number: MFCD00006690
CAS Number: 100-76-5
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR