Nonaethylene glycol 95%

Supplier: Ambeed
Warning

A482195-100MG A482195-10G A482195-1G A482195-250MG A482195-25G A482195-5G A482195-100G
76725-888EA 5 USD
76725-888 76725-890 76725-892 76725-894 76725-896 76725-898 77602-088
Nonaethylene glycol 95%
Nonaethylene glycol

Precaución: for Research Use Only

Formula: C₁₈H₃₈O₁₀
MDL Number: MFCD00698692
CAS Number: 3386-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR