n-Tricosane 99%

Supplier: Ambeed
Warning

Synonyms: Tricosane

A481517-100G A481517-1G A481517-25G A481517-5G
76725-880EA 593.05 USD
76725-880 76725-882 76725-884 76725-886
n-Tricosane 99%
n-Tricosane

Precaución: for Research Use Only

Formula: H₃C(CH₂)₂₁CH₃
MW: 324.63 g/mol
MDL Number: MFCD00009350
CAS Number: 638-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR