Pyrazolo[1,5-a]pyridin-2-ylmethanol 95%

Supplier: Ambeed
Warning

A480234-1G A480234-250MG
76725-872EA 325 USD
76725-872 77880-043
Pyrazolo[1,5-a]pyridin-2-ylmethanol 95%
Pyrazolo[1,5-a]pyridin-2-ylmethanol

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MDL Number: MFCD09878923
CAS Number: 76943-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR