N,N-Diisopropylaniline 97%

Supplier: Ambeed
Warning

A479161-1G A479161-250MG A479161-5G
76725-866EA 34 USD
76725-866 76725-868 76725-870
N,N-Diisopropylaniline 97%
N,N-Diisopropylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₉N
MDL Number: MFCD00048277
CAS Number: 4107-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR