Potassium (S)-2-hydroxypropanoate 60% in water

Supplier: Ambeed
Warning

A476988-100G A476988-25G
76725-844EA 13.8 USD
76725-844 76725-846
Potassium (S)-2-hydroxypropanoate 60% in water
Potassium (S)-2-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₃H₅KO₃
MW: 128.16 g/mol
MDL Number: MFCD00080809
CAS Number: 85895-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR