Isobutyl palmitate 98%

Supplier: Ambeed
Warning

Synonyms: Palmitic Acid Isobutyl Ester

A476242-100G A476242-25G A476242-500G A476242-5G
76725-836EA 226 USD
76725-836 76725-838 76725-840 76725-842
Isobutyl palmitate 98%
Isobutyl palmitate

Precaución: for Research Use Only

Formula: C₂₀H₄₀O₂
MDL Number: MFCD00059289
CAS Number: 110-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR