Methyl hesperidine 90% (by HPLC, sum of isomers)

Supplier: Ambeed
Warning

Synonyms: (S)-2-(3,4-Dimethoxyphenyl)-5-hydroxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one, Methyl-Hesperidin, Vitamin P (Water soluble)

A474885-5G A474885-100MG A474885-100G
76725-828EA 6 USD
76725-828 77669-138 77880-031
Methyl hesperidine 90% (by HPLC, sum of isomers)
Methyl hesperidine

Precaución: for Research Use Only

Formula: C₂₉H₃₆O₁₅
MDL Number: MFCD01741310
CAS Number: 11013-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR