N-Benzylacetamide 98%

Supplier: Ambeed
Warning

Synonyms: Lacosamide EP Impurity G, N-Acetylbenzylamine

A474806-100MG A474806-1G A474806-25G A474806-5G A474806-100G
76725-820EA 5.31 USD
76725-820 76725-822 76725-824 76725-826 77669-136
N-Benzylacetamide 98%
N-Benzylacetamide

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD00059204
CAS Number: 588-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR