(S)-1-tert-Butyl 2-methyl 1H-pyrrole-1,2(2H,5H)-dicarboxylate 95% stabilised

Supplier: Ambeed
Warning

Synonyms: N-BOC-3,4-didehydro-(S)-proline methyl ester, Boc-3,4-Dehydro-Pro-OMe

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A473143-1G A473143-250MG A473143-5G A473143-100MG
76725-804EA 36 USD
76725-804 76725-806 76725-808 77880-029
(S)-1-tert-Butyl 2-methyl 1H-pyrrole-1,2(2H,5H)-dicarboxylate 95% stabilised
(S)-1-tert-Butyl 2-methyl 1H-pyrrole-1,2(2H,5H)-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MW: 227.25 g/mol
MDL Number: MFCD00080341
CAS Number: 74844-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR