Potassium hexachloroplatinate(IV) 97%

Supplier: Ambeed
Danger

A472623-1G A472623-250MG A472623-5G
76725-800EA 54 USD
76725-800 76725-802 77880-028
Potassium hexachloroplatinate(IV) 97%
Potassium hexachloroplatinate(IV)

Precaución: for Research Use Only

Formula: K₂PtCl₆
MDL Number: MFCD00011389
CAS Number: 16921-30-5
UN: 3290
ADR: 6.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR