Neopentylamine 97%

Supplier: Ambeed
Danger

Synonyms: 1-Amino-2,2-dimethylpropane

A471405-5G A471405-25G
76725-796EA 25.47 USD
76725-796 77551-436
Neopentylamine 97%
Neopentylamine

Precaución: for Research Use Only

Formula: C₅H₁₃N
MW: 87.16 g/mol
MDL Number: MFCD00008134
CAS Number: 5813-64-9
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR