Propynol ethoxylate 90%

Supplier: Ambeed
Danger

Synonyms: Propargyl-PEG2-alcohol

A469411-25G A469411-5G
76725-782EA 11 USD
76725-782 76725-784
Propynol ethoxylate 90%
Propynol ethoxylate

Precaución: for Research Use Only

Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00099294
CAS Number: 3973-18-0
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR