Silver fluoride 99%

Supplier: Ambeed
Danger

A468128-10G A468128-25G A468128-1G A468128-5G
76725-768EA 74 USD
76725-768 76725-770 77551-424 77551-426
Silver fluoride 99%
Silver fluoride

Precaución: for Research Use Only

Formula: AgF
MDL Number: MFCD00003410
CAS Number: 7775-41-9
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR