EUK-134 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(SP-5-13)-Chloro[[2,2'-[1,2-ethanediylbis[(nitrilo-κN)methylidyne]]bis[6-methoxyphenolato-κO]](2-)]manganese, chloro[[2,2'-[1,2-ethanediylbis[(nitrilo-κN)methylidyne]]bis[6-methoxyphenolato-κO]]]-manganese
A466386-1MG
A466386-25MG
A466386-50MG
A466386-5MG
76725-750EA
16.98
USD
76725-750
76725-752
76725-754
76725-756
EUK-134 97%
EUK-134
Precaución: for Research Use Only
Formula:
C₁₈H₁₈ClMnN₂O₄ MW: 416.74 g/mol |
MDL Number:
MFCD08059577 CAS Number: 81065-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...