Boc-4-methylaminobenzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid

A466312-1G A466312-250MG
76725-748EA 81.69 USD
76725-748 77669-100
Boc-4-methylaminobenzoic acid 95%
Boc-4-methylaminobenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
MDL Number: MFCD04972273
CAS Number: 263021-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR