Methyl-4-hydroxybenzoate sodium salt 98%

Supplier: Ambeed
Warning

A465192-100G A465192-25G A465192-500G
76725-736EA 9 USD
76725-736 76725-738 76725-740
Methyl-4-hydroxybenzoate sodium salt 98%
Methyl-4-hydroxybenzoate sodium salt

Precaución: for Research Use Only

Formula: C₈H₇NaO₃
MDL Number: MFCD00016470
CAS Number: 5026-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR