Sorbitan monooleate Saponification Value 140-160

Supplier: Ambeed
Warning

A463047-100G A463047-25G A463047-500G
76725-718EA 6 USD
76725-718 76725-720 77669-082
Sorbitan monooleate Saponification Value 140-160
Sorbitan monooleate

Precaución: for Research Use Only

Formula: C₂₄H₄₄O₆
MDL Number: MFCD00080948
CAS Number: 1338-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR