Potassium methanesulfonate 98%

Supplier: Ambeed
Warning

A462310-1G A462310-25G A462310-5G
76725-712EA 10.61 USD
76725-712 76725-714 76725-716
Potassium methanesulfonate 98%
Potassium methanesulphonate

Precaución: for Research Use Only

Formula: CH₃KO₃S
MW: 134.2 g/mol
MDL Number: MFCD00070544
CAS Number: 2386-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR