2,2'-((Butane-1,4-diylbis(azanylylidene))bis(methanylylidene))diphenol 98%

Supplier: Ambeed
Warning

A461708-1G A461708-250MG A461708-5G
76725-706EA 47.75 USD
76725-706 76725-708 76725-710
2,2'-((Butane-1,4-diylbis(azanylylidene))bis(methanylylidene))diphenol 98%
2,2'-((Butane-1,4-diylbis(azanylylidene))bis(methanylylidene))diphenol

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂O₂
MW: 296.36 g/mol
MDL Number: MFCD00002246
CAS Number: 3955-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR