Hexyl-p-toluenesulfonate 95%

Supplier: Ambeed
Warning

Synonyms: p-Toluenesulphonic acid Hexyl Ester, Hexyl p-Toluenesulphonate

A460250-100G A460250-25G A460250-5G
76725-700EA 132.62 USD
76725-700 76725-702 76725-704
Hexyl-p-toluenesulfonate 95%
Hexyl-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₃H₂₀O₃S
MW: 256.36 g/mol
MDL Number: MFCD00059439
CAS Number: 3839-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR