S-Nitrosoglutathione (GSNO) 95%

Supplier: Ambeed
Danger

Synonyms: (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-(nitrosothio)-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, GSNO, Nitrosoglutathione, S-Nitroso-L-Glutathione

A460048-100MG A460048-250MG A460048-1G A460048-5G
76725-698EA 35 USD
76725-698 77601-994 77669-066 77872-760
S-Nitrosoglutathione (GSNO) 95%
S-Nitrosoglutathione (GSNO)

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₄O₇S
MW: 336.319 g/mol
MDL Number: MFCD00214341
CAS Number: 57564-91-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR