Pentaethylene glycol di-p-toluenesulfonate 98%

Supplier: Ambeed
Warning

A459002-1G A459002-5G A459002-25G A459002-100G
76725-678EA 18 USD
76725-678 76725-680 77601-992 77880-000
Pentaethylene glycol di-p-toluenesulfonate 98%
Pentaethylene glycol di-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₂₄H₃₄O₁₀S₂
MDL Number: MFCD00012204
CAS Number: 41024-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR