o-Terphenyl 98%

Supplier: Ambeed
Danger

A458457-25G A458457-5G
76725-664EA 12 USD
76725-664 76725-666
o-Terphenyl 98%
o-Terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₄
MDL Number: MFCD00003055
CAS Number: 84-15-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR