Dibutyl sebacate 98%

Supplier: Ambeed
Warning

Synonyms: Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate

A456299-100G A456299-25G A456299-1000G A456299-500G
76725-648EA 11 USD
76725-648 76725-650 77669-048 77669-050
Dibutyl sebacate 98%
Dibutyl sebacate

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₄
MW: 314.47 g/mol
Boiling Pt: 180
Melting Pt: -11
MDL Number: MFCD00027218
CAS Number: 109-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR