2-(Chloromethyl)-1-methylpiperidine hydrochloride 95%

Supplier: Ambeed
Danger

A456212-100MG A456212-1G A456212-250MG A456212-5G
76725-636EA 37.14 USD
76725-636 76725-638 76725-640 76725-642
2-(Chloromethyl)-1-methylpiperidine hydrochloride 95%
2-(Chloromethyl)-1-methylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅Cl₂N
MW: 184.11 g/mol
MDL Number: MFCD09800596
CAS Number: 27483-92-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR