Pentyl acrylate 98+% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with phenothiazine

A455665-5G A455665-1G
76725-626EA 69 USD
76725-626 77601-968
Pentyl acrylate 98+% stabilized
Pentyl acrylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MDL Number: MFCD00048845
CAS Number: 2998-23-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR