1-Butyl-3-methylpyridinium bromide 98%

Supplier: Ambeed
Warning

A455190-100G A455190-1G A455190-25G A455190-5G
76725-618EA 78 USD
76725-618 76725-620 76725-622 76725-624
1-Butyl-3-methylpyridinium bromide 98%
1-Butyl-3-methylpyridinium bromide

Precaución: for Research Use Only

Formula: C₁₀H₁₆BrN
MDL Number: MFCD01632121
CAS Number: 26576-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR