N-Methylisatoic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-benzo[d][1,3]oxazine-2,4-dione

A452941-10G A452941-1G A452941-25G A452941-5G A452941-100G
76725-590EA 82.76 USD
76725-590 76725-592 76725-594 76725-596 77669-034
N-Methylisatoic anhydride 98%
N-Methylisatoic anhydride

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.16 g/mol
MDL Number: MFCD00006815
CAS Number: 10328-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR