Methyl perfluoro-3,6-dioxadecanoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,2-difluoro-2-(1,1,2,2-tetrafluoro-2-(perfluorobutoxy)ethoxy)acetate

A451976-1G A451976-25G A451976-5G
76725-578EA 33.95 USD
76725-578 76725-580 76725-582
Methyl perfluoro-3,6-dioxadecanoate 98%
Methyl perfluoro-3,6-dioxadecanoate

Precaución: for Research Use Only

Formula: C₉H₃F₁₅O₄
MW: 460.10 g/mol
MDL Number: MFCD03094429
CAS Number: 129301-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR