Boc-Bpa-OH 98%

Supplier: Ambeed
Warning

Synonyms: Boc-p-Bz-Phe-OH

A450356-1G A450356-250MG
76725-566EA 65.78 USD
76725-566 76725-568
Boc-Bpa-OH 98%
Boc-Bpa-OH

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₅
MW: 369.417 g/mol
MDL Number: MFCD00151886
CAS Number: 104504-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR