N,N'-Bis(2-hydroxybenzyl)ethylenediamine 98%

Supplier: Ambeed
Warning

A448921-1G A448921-250MG A448921-5G
76725-554EA 39 USD
76725-554 76725-556 77601-946
N,N'-Bis(2-hydroxybenzyl)ethylenediamine 98%
N,N'-Bis(2-hydroxybenzyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MW: 272.3422 g/mol
MDL Number: MFCD00058922
CAS Number: 18653-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR