N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide 97%

Supplier: Ambeed
Warning

Synonyms: (Tosylazanediyl)bis(ethane-2,1-diyl) bis(4-methylbenzenesulphonate), N,N-Bis[2-(tosyloxy)ethyl]-p-toluenesulphonamide

A448081-25G A448081-5G
76725-544EA 33.95 USD
76725-544 76725-546
N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide 97%
N,N-Bis[2-(p-tolylsulfonyloxy)ethyl]-p-toluenesulfonamide

Precaución: for Research Use Only

Formula: C₂₅H₂₉NO₈S₃
MW: 567.69 g/mol
MDL Number: MFCD00026000
CAS Number: 16695-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR