2,6-Naphthalenedicarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Naphthalenedicarboxylic Acid, Naphthalene-2,6-dicarboxylic acid

A447213-100G A447213-10G A447213-1G A447213-250MG A447213-25G A447213-500G A447213-5G
76725-528EA 161.26 USD
76725-528 76725-530 76725-532 76725-534 76725-536 76725-538 76725-540
2,6-Naphthalenedicarboxylic acid 95%
2,6-Naphthalenedicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
MDL Number: MFCD00004105
CAS Number: 1141-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR